Skip to Content
Merck
All Photos(1)

Key Documents

779717

Sigma-Aldrich

19-Hydroxy-4-androstene-3,17-dione

≥90% (HPLC)

Synonym(s):

4-Androsten-19-ol-3,17-dione, 4-Androstene-3,17-dione-19-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26O3
CAS Number:
Molecular Weight:
302.41
Beilstein:
2567249
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

≥90% (HPLC)

form

crystals

optical activity

[α]/D +190±5°, c = 1 in chloroform

storage temp.

−20°C

SMILES string

C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34CO)[C@@H]1CCC2=O

InChI

1S/C19H26O3/c1-18-8-7-16-14(15(18)4-5-17(18)22)3-2-12-10-13(21)6-9-19(12,16)11-20/h10,14-16,20H,2-9,11H2,1H3/t14-,15-,16-,18-,19+/m0/s1

InChI key

XGUHPTGEXRHMQQ-BGJMDTOESA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J P Weniger et al.
Journal of steroid biochemistry, 35(3-4), 507-510 (1990-03-01)
Ovaries from 18-21-day-old foetal as well as from 2-10-day-old infantile rats were cultured in vitro in the presence of [3H]19-hydroxyandrostenedione and in the presence or absence of LH, FSH or (Bu)2 cAMP, and oestrone and oestradiol formed were determined by
M Minato et al.
Nihon Naibunpi Gakkai zasshi, 66(1), 29-41 (1990-01-20)
In order to evaluate the hypertensinogenic action of 19-hydroxyandrostenedione (19-OH-AD), which has been reported to be an amplifier of mineralocorticoid, the changes in several humoral factors were observed in 19-OH-AD treated rats as compared to those in DOCA hypertensive rats.
H Sekihara et al.
Clinical endocrinology, 39(5), 557-560 (1993-11-01)
We have reported that 19-hydroxyandrostenedione (19-OH-A-dione) functions as an amplifier of the sodium-retaining and hypertensive action of aldosterone. We therefore measured 19-hydroxyandrostenedione in hypertensive patients. We studied 53 normal male control subjects and 63 male patients with essential hypertension (48
D C Swinney et al.
Archives of biochemistry and biophysics, 305(1), 61-67 (1993-08-15)
Aromatase activity in microsomes from human placenta (HPM) and rat ovary (ROM) was compared by measuring the accumulation of 19-oxygenated intermediates, the effect of tritium substitution upon rates, and the distribution of tritium in products. A considerable accumulation of intermediates
Y Osawa et al.
Steroids, 55(4), 165-169 (1990-04-01)
19-Hydroxyandrostenedione (19-OHA) is secreted from the adrenal glands in men and women and also from the placenta during pregnancy. It has been found to cause hypertension in animal models. We have synthesized [7,7-2H2]-19-OHA with high deuterium content and, together with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service