Skip to Content
Merck
All Photos(1)

Documents

778990

Sigma-Aldrich

Sulfuric acid, fuming

reagent grade, 65.5-68.0% free SO3 basis (with NaOH)

Synonym(s):

Oleum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4 · (SO3)x
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 25 °C)

concentration

65.5-68.0% in NaOH (free SO3 )

density

1.925 g/mL at 25 °C (lit.)

SMILES string

O=S(=O)=O.OS(O)(=O)=O

InChI

1S/H2O4S.O3S/c1-5(2,3)4;1-4(2)3/h(H2,1,2,3,4);

InChI key

HIFJUMGIHIZEPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fuming sulfuric acid is an inorganic acid, produced by the addition of sulfur trioxide to concentrated sulfuric acid. It is commonly used for sulfonation reactions.1 Surface functionalization of poly (propylene) (PP) films using fuming sulfuric acid has been analyzed using X-ray photoelectron spectroscopy (XPS), attenuated total reflectance (ATR) infrared spectroscopy, and chemical modification. It is reported to assist the bromination of methylpyridines in the pyridine nucleus. Fuming sulfuric acid oxidizes fullerene C60 to form fullerene polymers.

Caution

may contain particle

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sulfonation of poly (propylene) films with fuming sulfuric acid.
Kaneko M and Sato H.
Macromolecular Chemistry and Physics, 206(4), 456-463 (2005)
Bromination of methylpyridines in fuming sulfuric acid.
Van der Does L and Hertog HJD.
Rec. Trav. Chim., 84(7), 951-964 (1965)
Products of reaction of fullerene C60 with fuming sulfuric acid studied by IR and ESR spectroscopy.
Garkusha OG, et al.
Russian Chemical Bulletin, 51(4), 628-631 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service