Skip to Content
Merck
All Photos(1)

Key Documents

778354

Sigma-Aldrich

Sodium SelenoSulfate Solution

Synonym(s):

Disodium selenosulfate, Selenosulfuric acid Disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
NA.23

composition

Sodium Selenosulfate, 8.2%
Sodium Sulfate, 12.8%
Water, 79%

Application

Sodium Selenosulfide is commonly used as a selenium source for the formation of nanoparticles in solution[1] including the formation of selenide quantum dots.[2] In many reactions it can serve the same purpose as selenourea but is more stable and easier to handle.[3]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Supplementary Hazards

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Zhu J, et al.
The Journal of Physical Chemistry B, 104(31), 7344-7347 (2000)
Song L, et al.
Catalysis Communications, 12(11), 1051-1054 (2011)
Pan D, et al.
The Journal of Physical Chemistry C, 111(15), 5661-5666 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service