Skip to Content
Merck
All Photos(1)

Key Documents

77787

Sigma-Aldrich

O-(2-Aminoethyl)-O′-(2-azidoethyl)nonaethylene glycol

≥90% (oligomer purity)

Synonym(s):

Azido-PEG-amine (n=10)

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$554.00

$554.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$554.00

About This Item

Empirical Formula (Hill Notation):
C22H46N4O10
CAS Number:
Molecular Weight:
526.62
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

$554.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥90% (oligomer purity)

form

liquid

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent

functional group

amine
azide

storage temp.

2-8°C

SMILES string

NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCN=[N+]=[N-]

InChI

1S/C22H46N4O10/c23-1-3-27-5-7-29-9-11-31-13-15-33-17-19-35-21-22-36-20-18-34-16-14-32-12-10-30-8-6-28-4-2-25-26-24/h1-23H2

InChI key

RMNAJNJBCBFOKX-UHFFFAOYSA-N

Application

O-(2-Aminoethyl)-O′-(2-azidoethyl)nonaethylene glycol may be used in the synthesis of potent PEGylated toll like receptor 7 (TLR7) ligand with improved solubility and modulated protein binding.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of PEGylated toll like receptor 7 ligands.
Chan M, et al.
Bioconjugate Chemistry, 22(3), 445-454 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service