Skip to Content
Merck
All Photos(1)

Key Documents

777129

Sigma-Aldrich

N,N-Diethylmethacrylamide

contains <500 ppm MEHQ as inhibitor, 97%

Synonym(s):

DEMAAM, DEMAM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO
CAS Number:
Molecular Weight:
141.21
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

contains

<500 ppm MEHQ as inhibitor

refractive index

n20/D 1.455

density

0.901 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCN(CC)C(=O)C(C)=C

InChI

1S/C8H15NO/c1-5-9(6-2)8(10)7(3)4/h3,5-6H2,1-2,4H3

InChI key

JMCVCHBBHPFWBF-UHFFFAOYSA-N

General description

Post polymerization, N, N-Diethyl methacrylamide gels show temperature sensitivity because of their lower critical solution temperature (LCST). This acrylamide is commonly polymerized to form temperature sensitive hydrogels. The presence of a hydrophobic group (methyl and ethyl group in this case) is characteristic in temperature sensitive polymers. Water solubility in such polymers decreases with increase in temperature.[1][2]

Application

Since the lower critical solution temperature (LCST) is around body temperature, it is widely used in the development of drug delivery systems based on the sol gel phase conversion at body temperature.[1][2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

183.0 °F

Flash Point(C)

83.88 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Drug release behavior of a pH/temperature sensitive calcium alginate/poly(N-acryloylglycine) bead with
core-shelled structure
Deng KL, et al.
Express Polymer Letters, 4(12), 773-780 (2010)
Y Qiu et al.
Advanced drug delivery reviews, 53(3), 321-339 (2001-12-18)
Environmentally sensitive hydrogels have enormous potential in various applications. Some environmental variables, such as low pH and elevated temperatures, are found in the body. For this reason, either pH-sensitive and/or temperature-sensitive hydrogels can be used for site-specific controlled drug delivery.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service