Skip to Content
Merck
All Photos(1)

Key Documents

777048

Sigma-Aldrich

2-[(7-{4-[N,N-Bis(4-methylphenyl)amino]phenyl}-2,1,3-benzothiadiazol-4-yl)methylene]propanedinitrile

97% (HPLC)

Synonym(s):

DTDCPB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H21N5S
CAS Number:
Molecular Weight:
483.59
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97% (HPLC)

form

powder

mp

285-290 °C

λmax

570 nm in dichloromethane

SMILES string

Cc1ccc(cc1)N(c2ccc(C)cc2)c3ccc(cc3)-c4ccc(\C=C(/C#N)C#N)c5nsnc45

InChI

1S/C30H21N5S/c1-20-3-10-25(11-4-20)35(26-12-5-21(2)6-13-26)27-14-7-23(8-15-27)28-16-9-24(17-22(18-31)19-32)29-30(28)34-36-33-29/h3-17H,1-2H3

InChI key

METIWNNPHPBEHP-UHFFFAOYSA-N

General description

2-((7-{4-[N,N-Bis(4-methylphenyl)amino]phenyl}-2,1,3-benzothiadiazol-4-yl)methylene)propanedinitrile (DTDCPB) is a donor-acceptor molecule which can be used in the fabrication of organic electronic devices such as organic solar cells (OSCs) and organic photovoltaics (OPVs).

Application

7.9% efficient vapor-deposited organic photovoltaic cells base on a simple bulk heterojunction

A vacuum-deposited organic solar cell employing this novel donor-acceptor-acceptor (D-A-A) donor molecule; DTDCPB; combined with the electron acceptor C60/ C70 achieved a record-high power conversion efficiency (PCE) of 6.8%.
Device structure:
MoO3 (30nm) / DTDCPB (7nm) / DTDCPB:C70 (40nm) / C70 (7nm) / BCP (10nm) / Ag (150nm)
Device performance:
  • JSC = 13.48 mA/cm2
  • VOC = 0.95 V
  • FF = 0.55
  • PCE = 6.8%

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

7.9% efficient vapor-deposited organic photovoltaic cells based on a simple bulk heterojunction
Zou Y, et al.
Journal of Material Chemistry A, 2(31), 12397-12402 (2014)
7.9% efficient vapor-deposited organic photovoltaic cells based on a simple bulk heterojunction
Yunlong Zou,
Journal of Material Chemistry A, 2, 12397-12402 (2014)
Effect of open-circuit voltage in organic solar cells based on various electron donor materials by inserting molybdenum trioxide anode buffer layer
Ke J, et al.
Solar Energy Materials and Solar Cells, 133(5), 248-254 (2015)
Vacuum-Deposited Small-Molecule Organic Solar Cells with High Power Conversion Efficiencies by Judicious Molecular Design and Device Optimization
Journal of the American Chemical Society, 134 (2012)
High fabrication yield organic tandem photovoltaics combining vacuum- and solution-processed subcells with 15% efficiency.
Xiaozhou C, et al.
Nature Energy, 3(5), 422-427 (2018)

Articles

Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service