Skip to Content
Merck
All Photos(1)

Key Documents

776688

Sigma-Aldrich

Gold nanorods

10 nm diameter, λ: 1064 nm Amax, dispersion in H2O

Synonym(s):

Au nanorod, gold nanorod, Au nanorods, Gold nanorod

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$986.00

$986.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$986.00

About This Item

MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

$986.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

colloidal suspension
dispersion in H2O
nanorod

contains

CTAB as stabilizer

concentration

>30 μg/mL in H2O

diam. × L

10 nm × 67 nm (±10%)

diameter

10 nm

density

1.00 g/mL at 25 °C

Mw/Mn

<10 (CV, monodispesity)

storage temp.

2-8°C

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

High aspect ratio nanorods allow the extension of the absorption peak into the IR; making them ideal for diagnostic applications in biological systems[1].

Gold Nanostructures: Properties and Applications
Gold Nanostructures: Properties and Applications

Legal Information

This product was produced under the methods claimed in U.S. Pat. Nos. 8,956,440 and 8,241,922

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prashant K Jain et al.
The journal of physical chemistry. B, 110(14), 7238-7248 (2006-04-08)
The selection of nanoparticles for achieving efficient contrast for biological and cell imaging applications, as well as for photothermal therapeutic applications, is based on the optical properties of the nanoparticles. We use Mie theory and discrete dipole approximation method to

Articles

Gold nanostructures such as nanorods, nanowires and microgold have found applications in exciting fields such as biomedical engineering, catalysis and diagnostics.

Silver nanomaterials have unique physical, chemical, and optical properties that are currently being leveraged for a wide variety of biological applications.

Among various ceramics, one-dimensional (1-D) piezoelectric ceramics have attracted significant scientific attention for use in energy harvesting.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service