Skip to Content
Merck
All Photos(1)

Key Documents

776491

Sigma-Aldrich

1H,1H-Perfluorooctyl methacrylate

contains MEHQ as inhibitor, 96%

Synonym(s):

1,1-Dihydroperfluorooctyl methacrylate, 1H,1H-Pentadecafluorooctyl methacrylate, 1H,H-Pentadecafluoro-1-octyl methacrylate, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-Pentadecafluoro-1-octyl methacrylate, Methacrylic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl ester, Nyebar

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$126.00
25 G
$402.00

$126.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$126.00
25 G
$402.00

About This Item

Empirical Formula (Hill Notation):
C12H7F15O2
CAS Number:
Molecular Weight:
468.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$126.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 30, 2025


Request a Bulk Order

Quality Level

Assay

96%

form

liquid

contains

MEHQ as inhibitor

density

1.574 g/mL at 25 °C

functional group

ester
fluoro
methacrylate

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C12H7F15O2/c1-4(2)5(28)29-3-6(13,14)7(15,16)8(17,18)9(19,20)10(21,22)11(23,24)12(25,26)27/h1,3H2,2H3

InChI key

RUEKTOVLVIXOHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

226.9 °F

Flash Point(C)

108.3 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service