Skip to Content
Merck
All Photos(1)

Key Documents

774561

Sigma-Aldrich

1-Methyl-1H-indole-5-carboxaldehyde

97%

Synonym(s):

1-Methylindolecarbaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$239.00

$239.00


Estimated to ship onApril 06, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$239.00

About This Item

Empirical Formula (Hill Notation):
C10H9NO
CAS Number:
Molecular Weight:
159.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$239.00


Estimated to ship onApril 06, 2025


Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

80-85 °C

functional group

aldehyde

storage temp.

2-8°C

SMILES string

[n]1(c2c(cc1)cc(cc2)C=O)C

InChI

1S/C10H9NO/c1-11-5-4-9-6-8(7-12)2-3-10(9)11/h2-7H,1H3

InChI key

XIVDZBIBWGQOTI-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niina Tani et al.
Bioorganic & medicinal chemistry, 22(23), 6655-6664 (2014-12-03)
Inhibition of CYP2A6-mediated nicotine metabolism can reduce cigarette smoking. We sought potent and selective CYP2A6 inhibitors to be used as leads for drugs useful in smoking reduction therapy, by evaluating CYP2A6 inhibitory effect of novel formyl, alkyl amine or carbonitrile

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service