Skip to Content
Merck
All Photos(1)

Documents

774049

Sigma-Aldrich

Zirconium(IV) oxide-yttria stabilized

sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.9% trace metals basis

Synonym(s):

8YSZ, YSZ8, YSZ, Yttria stabilized zirconia, Yttrium stabilized zirconium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZrO2 · Y2O3
CAS Number:
Molecular Weight:
349.03
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

solid

contains

8 mol % yttria as stabilizer

composition

(ZrO2)0.92(Y2O3)0.08

reaction suitability

core: zirconium

diam. × thickness

2.00 in. × 0.25 in.

mp

>2,600 °C (lit.)

SMILES string

O=[Zr]=O.O=[Y]O[Y]=O

InChI

1S/5O.2Y.Zr

InChI key

KSTOPEBVPBSOTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Solid Oxide Fuel cells operating at temperatures below 800 C (also known as intermediate temperature solid oxide fuel cell, IT-SOFC) are currently the topic of much research and development owing to the high degradation rates and materials costs incurred for SOFC operating at temperatures above 900 C. Thin films of electrode and electrolyte layers is one of the ways to achieve high performances in IT-SOFC. 8 mol% yttria stabilized zirconia sputtering targets can be used to deposit thin films of YSZ layers with high electronic conductivities.
Electrolyte material for solid oxide fuel cells (SOFC).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uhlenbruck, S.;
Solid State Ionics, 181, 447-447 (2010)
Lai, Bo-Kuai;
Journal of Power Sources, 196, 6299-6299 (2011)
Jee, Youngseok;
Journal of the Korean Ceramic Society, 47, 82-82 (2010)
Alexander B Tesler et al.
Nature communications, 6, 8649-8649 (2015-10-21)
Formation of unwanted deposits on steels during their interaction with liquids is an inherent problem that often leads to corrosion, biofouling and results in reduction in durability and function. Here we report a new route to form anti-fouling steel surfaces

Articles

Learn about the different low temperature direct alcohol fuel cells based on nanostructured material catalysts and anion exchange membrane fuel cells.

Thin film solid oxide fuel cells offer technical advantages; review of TF-SOFC deposition methods for electrolyte thin films.

This article describes the use and advantages of polyoxometalate-based redox-flow batteries as electrochemical energy storage systems over Li-ion batteries.

Nanocomposite Coatings with Tunable Properties Prepared by Atomic Layer Deposition

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service