Skip to Content
Merck
All Photos(1)

Key Documents

773980

Sigma-Aldrich

Zinc oxide

nanowires, size × L × 300 nm

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnO
CAS Number:
Molecular Weight:
81.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

nanowires

size L

× 300 nm

SMILES string

O=[Zn]

InChI

1S/O.Zn

InChI key

XLOMVQKBTHCTTD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Zinc oxide nanowires have been studied for use in UV Lasers; Solar Energy and Piezoelectrics.
Employed in the preparation of NaZnSiO3OH, a novel chiral framework material which has potential application in ion exchange, adsorption or catalysis.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhong Lin Wang et al.
Science (New York, N.Y.), 312(5771), 242-246 (2006-04-15)
We have converted nanoscale mechanical energy into electrical energy by means of piezoelectric zinc oxide nanowire (NW) arrays. The aligned NWs are deflected with a conductive atomic force microscope tip in contact mode. The coupling of piezoelectric and semiconducting properties
M H Huang et al.
Science (New York, N.Y.), 292(5523), 1897-1899 (2001-06-09)
Room-temperature ultraviolet lasing in semiconductor nanowire arrays has been demonstrated. The self-organized, <0001> oriented zinc oxide nanowires grown on sapphire substrates were synthesized with a simple vapor transport and condensation process. These wide band-gap semiconductor nanowires form natural laser cavities
Kurtis S Leschkies et al.
Nano letters, 7(6), 1793-1798 (2007-05-17)
We combine CdSe semiconductor nanocrystals (or quantum dots) and single-crystal ZnO nanowires to demonstrate a new type of quantum-dot-sensitized solar cell. An array of ZnO nanowires was grown vertically from a fluorine-doped tin oxide conducting substrate. CdSe quantum dots, capped
Jong-In Hahm
Journal of nanoscience and nanotechnology, 14(1), 475-486 (2014-04-16)
One-dimensional zinc oxide nanomaterials have been recently developed into novel, extremely effective, optical signal-enhancing bioplatforms. Their usefulness has been demonstrated in various biomedical fluorescence assays. Fluorescence is extensively used in biology and medicine as a sensitive and noninvasive detection method
Y Zhang et al.
Current molecular medicine, 13(10), 1633-1645 (2013-11-12)
Nanotechnology has witnessed tremendous advancement over the last several decades. Zinc oxide (ZnO), which can exhibit a wide variety of nanostructures, possesses unique semiconducting, optical, and piezoelectric properties hence has been investigated for a wide variety of applications. The most

Articles

Dye-sensitized solar cells (DSCs) are 3rd generation solar cells combining the promise of high efficiency with low production costs.

Organic photovoltaics (OPVs) represent a low-cost, lightweight, and scalable alternative to conventional solar cells. While significant progress has been made in the development of conventional bulk heterojunction cells, new approaches are required to achieve the performance and stability necessary to enable commercially successful OPVs.

Among various ceramics, one-dimensional (1-D) piezoelectric ceramics have attracted significant scientific attention for use in energy harvesting.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service