Skip to Content
Merck
All Photos(2)

Key Documents

773360

Sigma-Aldrich

Poly(2-ethyl-2-oxazoline) α-methyl, ω-2-hydroxyethylamine terminated

average Mn 2,000, PDI ≤1.2

Synonym(s):

POZ, Poly(2-ethyl-2-oxazoline) hydroxyethyl amine, Polyoxazoline, amino terminated polyoxazoline, aminoPEOx

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$230.00

$230.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$230.00

About This Item

Linear Formula:
CH3(C5H9NO)nNHC2H4OH
UNSPSC Code:
12162002
NACRES:
NA.23

$230.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

Tg 34°C

form

solid

mol wt

average Mn 2,000

PDI

≤1.2

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Amine polyoxazolines can be covalently attached through conjugation to carboxyl groups or sulfhydryl groups. Amines can also attach to oxidized carbohydrate groups (usually aldehydes) by reductive amination. Polyoxazolines are a biocompatible, hydrophilic, amorphous alternatives to poly(ethylene glycol).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Poly(2-oxazoline)s, synthesized via LCROP, resemble polypeptides and have various applications in polymer chemistry.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service