Skip to Content
Merck
All Photos(1)

Key Documents

772429

Sigma-Aldrich

α-Stn-Ser-ONa sodium salt

90%

Synonym(s):

O-[2-(Acetylamino)-6-O-(N-acetyl-α-neuraminosyl)-2-deoxy-α-D-galactopyranosyl]-L-serine disodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$313.00
25 MG
$1,090.00

$313.00


Only 0 left in stock

Request a Bulk Order

Select a Size

Change View
5 MG
$313.00
25 MG
$1,090.00

About This Item

Empirical Formula (Hill Notation):
C22H35N3Na2O16
Molecular Weight:
643.50
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

$313.00


Only 0 left in stock

Request a Bulk Order

Quality Level

Assay

90%

form

solid

optical activity

[α]22/D +62.0°, c = 0.2% in water

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H](CO[C@]2(C([O-])=O)C[C@H](O)[C@@H](NC(C)=O)[C@@]([C@H](O)[C@H](O)CO)([H])O2)O[C@H](OC[C@@H](C([O-])=O)N)[C@H](NC(C)=O)[C@H]1O.[Na+].[Na+]

InChI

1S/C22H37N3O16.2Na/c1-7(27)24-13-10(29)3-22(21(36)37,41-18(13)15(31)11(30)4-26)39-6-12-16(32)17(33)14(25-8(2)28)20(40-12)38-5-9(23)19(34)35;;/h9-18,20,26,29-33H,3-6,23H2,1-2H3,(H,24,27)(H,25,28)(H,34,35)(H,36,37);;/q;2*+1/p-2/t9-,10-,11+,12+,13+,14+,15+,16-,17+,18+,20-,22+;;/m0../s1

InChI key

HMONFCBAWSIFQQ-HKDDIGLCSA-L

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service