771929
Manganese iron oxide
nanopowder, 50 nm avg. part. size, 97% trace metals basis (BET)
Synonym(s):
Iron manganese oxide, Iron manganese spinel, Manganese ferrite
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
97% trace metals basis (BET)
form
nanopowder
avg. part. size
50 nm
mp
>300 °C
Application
Manganese Iron Oxide is an important nanoscale magnetic material with applications in the fabrication of magnetic nanoarrays and as an MRI contrast agent .
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Advanced Materials, 14, 231-234 (2002)
Biomaterials, 30(15), 2919-2928 (2009-02-24)
Iron oxide nanoparticles are effective contrast agents for enhancement of magnetic resonance imaging at tissue, cellular or even molecular levels. In this study, manganese doped superparamagnetic iron oxide (Mn-SPIO) nanoparticles were used to form ultrasensitive MRI contrast agents for liver
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service