Skip to Content
Merck
All Photos(1)

Key Documents

771031

Sigma-Aldrich

Cell Free Amino Acid Mixture-13C, 15N, D

2-100 mM in water, 98 atom % 13C, 97 atom % D, 98 atom % 15N

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,950.00

$1,950.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 EA
$1,950.00

About This Item

UNSPSC Code:
12352116
NACRES:
NA.12

$1,950.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

98 atom % 13C
98 atom % 15N
97 atom % D

Quality Level

form

liquid

concentration

2-100 mM in water

technique(s)

bio NMR: suitable
protein expression: suitable

shipped in

dry ice

storage temp.

−20°C

Related Categories

Application

This product suitable as source of Stable Isotope Labeled amino acids for Cell Free Protein Synthesis. For Mass Spec applications, consider product 909653, Stable Isotope Labeled Amino Acid Mixture for Mass Spec.

The product may appear turbid, due to the relatively high concentration of tyrosine. Once diluted for use, solution should be clear.

Packaging

1 mL aqueous solution of all 20 amino acids.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Analysis Note

Amino acid concentrations vary by lot. Please refer to the specific lot Certificate of Analysis.

Approximate concentrations (mM):
ASP 60
THR 35
SER 35
GLU 40
PRO 20
GLY 100
ALA 100
VAL 40
MET 10
ILE 30
LEU 45
TYR 10
PHE 16
HIS 5
LYS 15
ARG 10
GLN 20
ASN 20
TRP 20
CYS 20

required but not provided

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Filippo Caschera et al.
BioTechniques, 58(1), 40-43 (2015-01-22)
Here we present a procedure for preparing amino acid mixtures--having both the desired composition and a physiological pH--at high concentrations for cell-free expression systems. Up to 2.1 mg/mL of active protein was synthesized in batch mode reactions with an all

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service