Skip to Content
Merck
All Photos(1)

Key Documents

769525

Sigma-Aldrich

Yttrium fluoride

granular, ≤1 mm

Synonym(s):

Yttrium(III) fluoride

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$246.00

$246.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$246.00

About This Item

Linear Formula:
YF3
CAS Number:
Molecular Weight:
145.90
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23
grade:
for analytical purposes
form:
granular

$246.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

Quality Level

form

granular

impurities

≤0.1% PBB (polybrominated biphenyls)
≤0.1% PBDE (polybrominated diphenyl ethers)
≤0.2% O (Oxygen)

particle size

≤1 mm
<1 mm, coeff var ≥90%

density

4.01 g/mL at 25 °C (lit.)

cation traces

Cd: ≤0.01%
Co: ≤0.001%
Cr: ≤0.001%
Cu: ≤0.001%
Fe: ≤0.002%
Hg: ≤0.1%
Mn: ≤0.001%
Pb: ≤0.1%

SMILES string

F[Y](F)F

InChI

1S/3FH.Y/h3*1H;/q;;;+3/p-3

InChI key

RBORBHYCVONNJH-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Fluorides of rare earths find applications in anti-reflective coatings. They are also used in synthesis of luminescent bio-imaging probes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dual-Modality in Vivo Imaging Using Rare-Earth Nanocrystals With Near-Infrared to Near-Infrared (NIR-to-NIR) Upconversion Luminescence and Magnetic Resonance Properties
Zhou, J., et al.
Biomaterials, 31(12), 3287-3295 (2010)
Chemical processing for inorganic fluoride and oxyfluoride materials having optical functions
Fujihara, S. and Tokumo, K.
Journal of Fluorine Chemistry, 130(12), 1106-1110 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service