Skip to Content
Merck
All Photos(2)

Documents

76830

Sigma-Aldrich

3,3,6,9,9-Pentamethyl-2,10-diazabicyclo[4.4.0]dec-1-ene

≥96.0% (GC)

Synonym(s):

1,2,3,4,4a,5,6,7-Octahydro-2,2,4a,7,7-pentamethylnaphthyridine, PMDBD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H24N2
CAS Number:
Molecular Weight:
208.34
Beilstein:
779556
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.0% (GC)

form

liquid

density

0.924 g/mL at 20 °C (lit.)

SMILES string

CC1(C)CCC2(C)CCC(C)(C)N=C2N1

InChI

1S/C13H24N2/c1-11(2)6-8-13(5)9-7-12(3,4)15-10(13)14-11/h6-9H2,1-5H3,(H,14,15)

InChI key

OEOPGVPCZRQSMQ-UHFFFAOYSA-N

Other Notes

Amidine base with exceptional properties

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P.H. Boyle et al.
Journal of the Chemical Society. Chemical Communications, 239-239 (1992)
S.E. Denmark
The Journal of Organic Chemistry, 46, 3144-3144 (1981)
A. Eschenmoser et al.
Angewandte Chemie (International Edition in English), 91, 670-670 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service