Skip to Content
Merck
All Photos(1)

Key Documents

767735

Sigma-Aldrich

9,9′-Spirobifluorene

97%

Synonym(s):

9,9′-Spirobi[9H-fluorene]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H16
CAS Number:
Molecular Weight:
316.39
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

195-200 °C

SMILES string

c1ccc2c(c1)-c3ccccc3C24c5ccccc5-c6ccccc46

InChI

1S/C25H16/c1-5-13-21-17(9-1)18-10-2-6-14-22(18)25(21)23-15-7-3-11-19(23)20-12-4-8-16-24(20)25/h1-16H

InChI key

SNFCXVRWFNAHQX-UHFFFAOYSA-N

General description

9,9′-Spirobifluorene is a polyfluorene with carbon atoms of the methylene bridge connected to two fluorene molecules. It is mainly used in organic electronics due to its robust structure and two perpendicularly arranged π systems.

Application

9,9′-Spirobifluorene can be used in the synthesis of spirobifluorene based conjugated microporous polymers which can be potentially be used as a gas absorbent. It may be used as a hole transporting material which can be used in the fabrication of perovskite solar cells.

Features and Benefits

High glass-transition temperature, excellent thermal stability, soluble in common organic solvents, multi-layer devices can be fabricated by thermal cross-linking of cast films. Easy colour tunability through the introduction of low band gap co-monomers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Anodic oxidation of 9, 9?-spirobifluorene in CH2Cl2+ 0.2 M Bu4NBF4. Electrochemical behaviour of the derived oxidation product.
Rault-Berthelot J, et al.
Synthetic Metals, 97(3), 211-215 (1998)
Organic Light Emitting Diode (OLED) materials from spirobifluorene derivatives
R. N. Jana, S. Chakraborty, R. Banerjee
Physics Express, 4(2) (2014)
Photocatalytic H2 Evolution by Pt-Loaded 9, 9?-Spirobifluorene-Based Conjugated Microporous Polymers under Visible-Light Irradiation.
Modak A, et al.
Bulletin of the Chemical Society of Japan, 89(8), 887-891 (2016)
Amorphous poly-2, 7-fluorene networks
Marsitzky, D., Murray, J., Scott, J. C., & Carter, K. R.
Chemistry of Materials, 13(11), 4285-4289 (2001)
Synthesis of 9, 9?-spirobifluorene-based conjugated microporous polymers by FeCl 3-mediated polymerization.
Modak A, et al.
Polym. Chem., 7(6), 1290-1296 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service