Skip to Content
Merck
All Photos(1)

Key Documents

766518

Sigma-Aldrich

2,2,2-Trifluoroethoxyamine hydrochloride

Synonym(s):

O-(2,2,2-Trifluoroethyl)hydroxylamine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$117.00

$117.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$117.00

About This Item

Empirical Formula (Hill Notation):
C2H5ClF3NO
CAS Number:
Molecular Weight:
151.52
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$117.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

form

solid

mp

161-163 °C

functional group

fluoro

storage temp.

2-8°C

SMILES string

NOCC(F)(F)F.Cl

InChI

1S/C2H4F3NO.ClH/c3-2(4,5)1-7-6;/h1,6H2;1H

InChI key

MIQJCHKNVAHUGJ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service