Skip to Content
Merck
All Photos(1)

Key Documents

766267

Sigma-Aldrich

Chlorotris(triphenylphosphine)ruthenium(II) acetate

97%

Synonym(s):

(Acetato)chlorotris(triphenylphosphine)ruthenium(II), RuCl(OAc)(PPh3)3

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
$287.00

$287.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2 G
$287.00

About This Item

Empirical Formula (Hill Notation):
C56H48ClO2P3Ru
CAS Number:
Molecular Weight:
982.42
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$287.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst

mp

223-244 °C

SMILES string

CC(=O)O[Ru]Cl.c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6.c7ccc(cc7)P(c8ccccc8)c9ccccc9

InChI

1S/3C18H15P.C2H4O2.ClH.Ru/c3*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-2(3)4;;/h3*1-15H;1H3,(H,3,4);1H;/q;;;;;+2/p-2

InChI key

CBECNVOKQNQZIN-UHFFFAOYSA-L

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service