Skip to Content
Merck
All Photos(1)

Key Documents

766054

Sigma-Aldrich

P-Chlorodiphenylphosphine

99%

Synonym(s):

Diphenylphosphinous chloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$132.00

$132.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$132.00

About This Item

Linear Formula:
(C6H5)2PCl
CAS Number:
Molecular Weight:
220.63
Beilstein:
512032
EC Number:
MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22

$132.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

vapor density

7.8 (vs air)

Assay

99%

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.636 (lit.)

bp

320 °C (lit.)

density

1.194 g/mL at 25 °C
1.229 g/mL at 25 °C (lit.)

functional group

phosphine

SMILES string

ClP(c1ccccc1)c2ccccc2

InChI

1S/C12H10ClP/c13-14(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H

InChI key

XGRJZXREYAXTGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

280.4 °F

Flash Point(C)

138 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jaewoo Kwak et al.
Dalton transactions (Cambridge, England : 2003), 44(10), 4765-4772 (2015-02-11)
A series of mono-, di-, and tri-phosphine oxide substituted triarylboranes, Mes2BAr (1), MesBAr2 (2), and BAr3 (3) (Ar = 4-(Ph2PO)-2,6-Me2-C6H2) were prepared to investigate the effect of a phosphine oxide group (Ph2PO) on Lewis acidity enhancement of triarylboranes. The X-ray

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service