Skip to Content
Merck
All Photos(1)

Key Documents

765473

Sigma-Aldrich

Gold nanoparticles

15 nm diameter, carboxylic acid functionalized, PEG 3000 coated, OD 50, dispersion in H2O

Synonym(s):

Gold nanoparticles COOH functionalized, Au NP COOH, Gold Colloid

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$534.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$534.00

About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

$534.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

material

PEG 3000

form

dispersion in H2O
nanoparticles

packaging

glass bottle of 1 mL

OD

50

diameter

15 nm

pH

6.0-8.0 (25 °C)

bp

100 °C (lit.)

solubility

water: miscible

density

1.00 g/cm3

λmax

520 nm

functional group

carboxylic acid

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Carboxylic acid functionalized GNPs form conjugates with various protein molecules due to the electrostatic attraction between negatively charged carboxylate groups and positively charged amino groups of protein.[1]

Application

Functionalized Particles have been extensively used to study cellular uptake of nanoparticles and targeted drug delivery.
The carboxylic acid group binds with various proteins and is used in targeted drug delivery and gene therapy. [2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nishima Wangoo et al.
Journal of colloid and interface science, 323(2), 247-254 (2008-05-20)
We report a novel strategy for the synthesis of aqueous stable, carboxylated gold nanoparticles (GNPs) by using glutamic acid as the reducing agent. The ratio of chloroaurate ions, AuCl(-)(4) to glutamic acid was optimized in the reaction medium to obtain
Functionalized gold nanoparticles and their biomedical applications
Tiwari, P. M., Vig, K., Dennis, V. A., & Singh, S. R.
Nanomaterials, 1(1), 31-63 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service