Skip to Content
Merck
All Photos(1)

Key Documents

764981

Sigma-Aldrich

9,10-Bis(2-naphthyl)anthrace

99% (HPLC)

Synonym(s):

9,10-Di-2-naphthalenyl-anthracene, ADN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H22
CAS Number:
Molecular Weight:
430.54
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99% (HPLC)

form

solid

mp

383-387 °C

SMILES string

c1ccc2cc(ccc2c1)-c3c4ccccc4c(-c5ccc6ccccc6c5)c7ccccc37

InChI

1S/C34H22/c1-3-11-25-21-27(19-17-23(25)9-1)33-29-13-5-7-15-31(29)34(32-16-8-6-14-30(32)33)28-20-18-24-10-2-4-12-26(24)22-28/h1-22H

InChI key

VIZUPBYFLORCRA-UHFFFAOYSA-N

Application

Fluorescent host material in high efficiency pure blue devices.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lu-Qi Wang et al.
Oncotarget, 6(42), 44108-44122 (2015-12-18)
Ketamine enhances autonomic activity, and unmyelinated C-type baroreceptor afferents are more susceptible to be blocked by ketamine than myelinated A-types. However, the presynaptic transmission block in low-threshold and sex-specific myelinated Ah-type baroreceptor neurons (BRNs) is not elucidated. Action potentials (APs)
Juan Sanchis et al.
Revista espanola de cardiologia (English ed.), 72(4), 317-323 (2018-04-16)
Cell-free DNA (cfDNA) in ST-segment elevation myocardial infarction might originate from hyperactivated leukocytes at the coronary lesion. Our aim was to investigate the relationship between cfDNA and coronary reperfusion. We studied 116 patients treated with primary angioplasty using thrombus aspiration.
María Valeria Rumi et al.
Veterinary microbiology, 230, 228-234 (2019-03-05)
Extended-spectrum β-lactamase (ESBL), plasmid-mediated AmpC (pAmpC) and MCR-1 phosphoethanolamine transferase enzymes have been pointed out as the main plasmid-mediated mechanisms of resistance to third generation cephalosporins (TGC) and colistin, respectively, and are currently considered a major concern both in human
Barbara Rieck et al.
PLoS pathogens, 13(5), e1006399-e1006399 (2017-05-26)
Sensing and response to changes in nutrient availability are essential for the lifestyle of environmental and pathogenic bacteria. Serine/threonine protein kinase G (PknG) is required for virulence of the human pathogen Mycobacterium tuberculosis, and its putative substrate GarA regulates the
Douaa Farhat et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 18(10), 1333-1344 (2017-02-24)
Recently, dinitriles (NC(CH

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service