Skip to Content
Merck
All Photos(3)

Key Documents

763691

Sigma-Aldrich

Lanthanum strontium cobaltite, LSC-82

greener alternative

99% trace metals basis

Synonym(s):

Cobalt lanthanum strontium oxide, LSC, LSC-82, LSCo, Lanthanum strontium cobalt oxide, Strontium doped lanthanum cobalt oxide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$332.00

$332.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$332.00

About This Item

Empirical Formula (Hill Notation):
La0.8Sr0.2CoO3
CAS Number:
Molecular Weight:
235.58
MDL number:
UNSPSC Code:
26111700
NACRES:
NA.23

$332.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99% trace metals basis

form

powder

mol wt

Mw 235.58 g/mol

composition

La0.8Sr0.2CoO3

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

surface area

13-18 m2/g

particle size

0.4-1 μm (Horiba-Laser)

application(s)

battery manufacturing

greener alternative category

General description

LSC-82 is a promising cathode material for Solid Oxide Fuel Cells with a pervoskite type structure.
LSC-82 has the unique ability to reduce itself reversibly at moderate partial pressures of oxygen, thus producing large numbers of oxygen ion vacancies. The ion vacancies allow a number of events to take place, such as;
  • ease the transport of oxygen ion and cause oxygen surface exchange,
  • raise mechanical stress by causing volume expansion of the lattice.
Influence of change in temperature on the properties of LSC-82 has been reported in detail.[1] Effect of temperature on magnetic susceptibility has also been reported.[2]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

SOFCs containing LSC-82 cathodes show good performances with doped ceria electrolytes compared to yttria stabilized zirconia electrolytes.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High Temperature Magnetic Properties of Sr-Doped Lanthanum Cobalt Oxide (La1- x Sr x CoO3- δ).
Bhaskar D, et al.
Chemistry of Materials, 20(8), 2624-2628 null
Bouwmeester, H. J. M.; et al.
Solid State Ionics, 72, 185-185 (1994)
Milind J Gadre et al.
Physical chemistry chemical physics : PCCP, 14(8), 2606-2616 (2012-01-25)
An interface between the perovskite La(0.8)Sr(0.2)CoO(3-δ) (LSC-113) and the K(2)NiF(4)-type (La(0.5)Sr(0.5))(2)CoO(4-δ) (LSC-214) heterostructure was recently shown to enhance oxygen surface exchange and the rate of the oxygen reduction reaction (ORR) by orders of magnitude compared to either the LSC-113 or
Thermal and chemical expansion of Sr-doped lanthanum cobalt oxide (La1-x Sr x CoO3-δ).
Chen X, et al.
Chemistry of Materials, 17(17), 4537-4546 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service