Skip to Content
Merck
All Photos(1)

Key Documents

762857

Sigma-Aldrich

Poly(N-isopropylacrylamide-co-butylacrylate)

butylacrylate 5 mol %, average Mn 30,000

Synonym(s):

NIPAM, polyNIPAM, polyNIPAM-BA

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$1,080.00

$1,080.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$1,080.00

About This Item

Linear Formula:
(C6H11NO)m(C7H12O2)n
UNSPSC Code:
12162002
NACRES:
NA.23

$1,080.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

feed ratio

NIPAM:BA 20:1

mol wt

average Mn 30,000

composition

butylacrylate, 5 mol %

transition temp

LCST 25 °C
Tm 345-351

Looking for similar products? Visit Product Comparison Guide

Application

This copolymer shows LCST at 25°C (lower temperature than PolyNIPAM), which makes it useful in hydrogel and tissue engineering applications needing LCST close to room temperature or below.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

By altering the physicochemical properties, smart or intelligent drug delivery systems can be designed to deliver therapeutic molecules on-demand. Learn more about the application of stimuli-responsive materials in drug delivery.

Immunosuppressive tumor-associated myeloid cells (TAMC) are responsible for glioblastoma (GBM) resistance to immunotherapies and existing standard of care treatments. This mini-review highlights recent progress in implementing nanotechnology in advancing TAMC-targeted therapies for GBM.

Biomaterials science integrates smart materials into biological research, requiring a deep understanding of biological systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service