75986
Chlorotriethylsilane
Wacker Chemie AG, ≥99.0%
Synonym(s):
Silane E3, TESCl, Triethylchlorosilane
Select a Size
Select a Size
About This Item
Recommended Products
Assay
≥99.0%
form
liquid
manufacturer/tradename
Wacker Chemie AG
refractive index
n20/D 1.43 (lit.)
bp
142-144 °C (lit.)
density
0.898 g/mL at 25 °C (lit.)
SMILES string
CC[Si](Cl)(CC)CC
InChI
1S/C6H15ClSi/c1-4-8(7,5-2)6-3/h4-6H2,1-3H3
InChI key
DCFKHNIGBAHNSS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Other Notes
comparable product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
86.0 °F
Flash Point(C)
30 °C
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service