758523
4-Bromo-2-(trifluoromethyl)pyridine hydrobromide
95%
About This Item
Recommended Products
Assay
95%
form
solid
mp
96-101 °C
storage temp.
−20°C
SMILES string
Br.FC(F)(F)c1cc(Br)ccn1
InChI
1S/C6H3BrF3N.BrH/c7-4-1-2-11-5(3-4)6(8,9)10;/h1-3H;1H
InChI key
XVEXNWJBZDKDBQ-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service