Skip to Content
Merck
All Photos(2)

Key Documents

757373

Sigma-Aldrich

Tantalum(V) oxide

tablet, deposition grade, 99.99% trace metals basis

Synonym(s):

Tantalum pentoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ta2O5
CAS Number:
Molecular Weight:
441.89
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

grade

deposition grade

Assay

99.99% trace metals basis

form

tablet

density

8.2 g/mL at 25 °C (lit.)

SMILES string

O=[Ta](=O)O[Ta](=O)=O

InChI

1S/5O.2Ta

InChI key

PBCFLUZVCVVTBY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu, L.; et al.
Green Chemistry, 10, 746-746 (2008)
Schmitt, K.; et al.
Sensors, 8, 711-711 (2008)
Kao, C.-H.; et al.
Applied Physics Letters, 96, 112901-112901 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service