Skip to Content
Merck
All Photos(2)

Key Documents

756563

Sigma-Aldrich

Sodium yttrium fluoride, ytterbium and thulium doped

99% trace metals basis

Synonym(s):

Blue up-conversion phosphor, Sodium thulium ytterbium yttrium fluoride, Ytterbium and thulium doped sodium yttrium fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
NaY0.77Yb0.20Tm0.03F4
Molecular Weight:
207.12
UNSPSC Code:
12352300

Assay

99% trace metals basis

form

powder

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kramer, K.M.; et al.
Chemistry of Materials, 16, 1244-1244 (2004)
Yang, X.; et al.
J. Mater. Sci., 42, 7042-7042 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service