Skip to Content
Merck
All Photos(2)

Documents

756415

Sigma-Aldrich

Calcium nitride

powder, -200 mesh, 99% trace metals basis (contains <0.5% Mg)

Synonym(s):

Tricalcium dinitride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca3N2
CAS Number:
Molecular Weight:
148.25
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99% trace metals basis (contains <0.5% Mg)

form

powder

particle size

-200 mesh

density

2.63 g/mL at 25 °C (lit.)

cation traces

Mg: <0.5%

SMILES string

[Ca]=N[Ca]N=[Ca]

InChI

1S/3Ca.2N

InChI key

DVJMVZQXJNJSJR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Calcium nitride is used as a nitride source in metathesis reactions to produce complex nitrides as well as SiAlON-related optical materials.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hampshire, S.; et al.
Nature, 274, 880-880 (1978)
Piao, X.; et al.
Chemistry of Materials, 19, 4592-4592 (2007)
Chern, M.Y.; Vennos, D.A.; DiSalvo, F.J.
Journal of Solid State Chemistry, 96, 415-415 (1992)

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service