Skip to Content
Merck
All Photos(1)

Key Documents

756369

Sigma-Aldrich

1,3-Dicyclohexylimidazolium chloride

Synonym(s):

1,3-Dicyclohexyl-1H-imidazolium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$239.00
5 G
$811.00

$239.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$239.00
5 G
$811.00

About This Item

Empirical Formula (Hill Notation):
C15H25ClN2
CAS Number:
Molecular Weight:
268.83
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$239.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

reaction suitability

reagent type: catalyst

mp

101-144 °C (decomposition)

SMILES string

[Cl-].C1CCC(CC1)n2cc[n+](c2)C3CCCCC3

InChI

1S/C15H25N2.ClH/c1-3-7-14(8-4-1)16-11-12-17(13-16)15-9-5-2-6-10-15;/h11-15H,1-10H2;1H/q+1;/p-1

InChI key

UPJKDKOHHMKJAY-UHFFFAOYSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rimesh Augustine et al.
Journal of materials chemistry. B, 8(26), 5745-5755 (2020-06-11)
A series of multi-stimuli-responsive poly(N-isopropylacrylamide)30-SS-block-poly(L-lysine)30-block-poly(caprolactone)n (p(NIPAM)30-SS-b-p(Lys)30-b-p(CL)n) (n = 50, 75, 100, 125) triblock copolymers have been synthesized by combining reversible addition-fragmentation chain transfer polymerization of NIPAM, organo-catalyzed ring-opening polymerization (ROP) of Z-lysine N-carboxyanhydride and metal-catalyzed ROP of CL with an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service