Skip to Content
Merck
All Photos(1)

Key Documents

755508

Sigma-Aldrich

8-Methylquinoline N-oxide

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$160.00
5 G
$763.00

$160.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$160.00
5 G
$763.00

About This Item

Empirical Formula (Hill Notation):
C10H9NO
CAS Number:
Molecular Weight:
159.18
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$160.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

mp

240-260 °C

storage temp.

2-8°C

SMILES string

[O-][N+]1=CC=CC2=C1C(C)=CC=C2

InChI

1S/C10H9NO/c1-8-4-2-5-9-6-3-7-11(12)10(8)9/h2-7H,1H3

InChI key

PIOGDSHYODXWAK-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biao Lu et al.
Journal of the American Chemical Society, 132(40), 14070-14072 (2010-09-22)
Gold-catalyzed intermolecular oxidations of internal alkynes have been achieved with high regioselectivities using 8-alkylquinoline N-oxides as oxidants and in the absence of acid additives. Synthetically versatile α,β-unsaturated carbonyls are obtained in good to excellent yields and with excellent E-selectivities. A

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service