Skip to Content
Merck
All Photos(1)

Key Documents

754080

Sigma-Aldrich

ADT

sublimed, 97%

Synonym(s):

Anthra[2,3-b:6,7-b′]dithiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H10S2
CAS Number:
Molecular Weight:
290.40
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

form

sublimed

loss

0.5 wt. %, 280 °C

semiconductor properties

P-type (mobility=0.3 cm2/V·s)

SMILES string

c1cc2cc3cc4cc5sccc5cc4cc3cc2s1

InChI

1S/C18H10S2/c1-3-19-17-9-15-8-14-6-12-2-4-20-18(12)10-16(14)7-13(15)5-11(1)17/h1-10H

InChI key

DAMUWSYTQPWFIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

ADT is an anthradithiophene based small molecule semiconductor that contributes to a new class of heteroacenes, which has a 22-electron π-conjugated system. It can be majorly used in a variety of organic electronic applications.[1][2]

Application

ADT can be used as a conducting polymer in the fabrication of organic field effect transistors (OFETs) and organic photovoltaics (OPVs).[3][4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Physical Properties of Anthra [2, 3-b; 6, 7-b'] Dithiophene Organic Material
Hamad H
Journal of Physical Chemistry and Functional Materials, 2(1), 37-39 (2019)
Synthesis of 2, 2?-[2, 2?-(arenediyl) bis (anthra [2, 3-b] thiophene-5, 10-diylidene)] tetrapropanedinitriles and their performance as non-fullerene acceptors in organic photovoltaics
Baranov DS, et al.
Synthetic Metals, 255(18), 116097-116097 (2019)
Isomerically pure anthra [2, 3-b: 6, 7-b?]-difuran (anti-ADF),-dithiophene (anti-ADT), and-diselenophene (anti-ADS): selective synthesis, electronic structures, and application to organic field-effect transistors
Nakano M, et al.
The Journal of Organic Chemistry, 77(18), 8099-8111 (2012)
Bogdana Borca et al.
ACS nano, 9(12), 12506-12512 (2015-11-19)
Single molecular switches are basic device elements in organic electronics. The pentacene analogue anthradithiophene (ADT) shows a fully reversible binary switching between different adsorption conformations on a metallic surface accompanied by a charge transfer. These transitions are activated locally in
Florence K Keane et al.
Cancer, 121(16), 2713-2719 (2015-05-01)
Radiotherapy (RT), short-course androgen deprivation therapy (ADT), and brachytherapy in various combinations are treatment options for patients with intermediate-risk prostate cancer (PC), but the question of which combination if any is necessary to minimize PC-specific mortality (PCSM) risk in patients

Articles

Organic materials in optoelectronic devices like LEDs and solar cells are of significant academic and commercial interest.

Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.

Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service