Skip to Content
Merck
All Photos(1)

Key Documents

753572

Sigma-Aldrich

Sodium sulfite-34S

90 atom % 34S, 95% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$1,850.00

$1,850.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
10 MG
$1,850.00

About This Item

Linear Formula:
Na234SO3
Molecular Weight:
127.95
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.12

$1,850.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

90 atom % 34S

Quality Level

Assay

95% (CP)

form

solid

mass shift

M+2

storage temp.

room temp

SMILES string

[Na+].[Na+].[O-][34S]([O-])=O

InChI

1S/2Na.H2O3S/c;;1-4(2)3/h;;(H2,1,2,3)/q2*+1;/p-2/i;;4+2

InChI key

GEHJYWRUCIMESM-FRWSWXKJSA-L

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaojun Zhang et al.
Journal of agricultural and food chemistry, 63(12), 3129-3134 (2015-03-11)
A highly selective and sensitive method was developed for the determination of tetrodotoxin (TTX) in marine organisms by immunoaffinity chromatography (IAC) purification coupled with ultrahigh performance liquid chromatography tandem mass spectrometry (UPLC-MS/MS). An IAC column was prepared and used to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service