Skip to Content
Merck
All Photos(1)

Key Documents

751294

Sigma-Aldrich

4-Oxocyclohexanecarboxylic acid

97%

Synonym(s):

4-Ketocyclohexanecarboxylic acid, Cyclohexanone-4-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$70.70

$70.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$70.70

About This Item

Empirical Formula (Hill Notation):
C7H10O3
CAS Number:
Molecular Weight:
142.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$70.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

67-71 °C

functional group

carboxylic acid
ketone

storage temp.

2-8°C

SMILES string

OC(=O)C1CCC(=O)CC1

InChI

1S/C7H10O3/c8-6-3-1-5(2-4-6)7(9)10/h5H,1-4H2,(H,9,10)

InChI key

OWLXUYGCLDGHJJ-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Kaneda et al.
European journal of biochemistry, 218(3), 997-1003 (1993-12-15)
We have previously demonstrated that Corynebacterium cyclohexanicum degrades cyclohexanecarboxylic acid, a bacteriocide, through a pathway including the aromatization of 4-oxocyclohexanecarboxylic acid to 4-hydroxybenzoic acid [Kaneda, T. (1974) Biochem. Biophys. Res. Commun. 58, 140-144]. Aromatization has now been shown to be

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service