Skip to Content
Merck
All Photos(1)

Key Documents

748900

Sigma-Aldrich

1,9-Nonanedithiol

99%

Synonym(s):

1,9-Dimercaptononane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HS(CH2)9SH
CAS Number:
Molecular Weight:
192.39
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

liquid

refractive index

n20/D 1.4999 (lit.)
n20/D 1.501

bp

284 °C (lit.)

density

0.939 g/mL at 25 °C
0.952 g/mL at 25 °C (lit.)

SMILES string

SCCCCCCCCCS

InChI

1S/C9H20S2/c10-8-6-4-2-1-3-5-7-9-11/h10-11H,1-9H2

InChI key

GJRCLMJHPWCJEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Wei Zhao et al.
Small (Weinheim an der Bergstrasse, Germany), 11(35), 4509-4516 (2015-06-04)
The ability to tune gauge factors in terms of magnitude and orientation is important for wearable and conformal electronics. Herein, a sensor device is described which is fabricated by assembling and printing molecularly linked thin films of gold nanoparticles on

Articles

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service