Skip to Content
Merck
All Photos(1)

Key Documents

747629

Sigma-Aldrich

Cu(dap)2 chloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$127.00
100 MG
$211.00

$127.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$127.00
100 MG
$211.00

About This Item

Empirical Formula (Hill Notation):
C52H40ClCuN4O4
Molecular Weight:
883.90
UNSPSC Code:
12161600
NACRES:
NA.22

$127.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

form

powder

Quality Level

reaction suitability

core: copper
reaction type: Photocatalysis
reagent type: catalyst

mp

160-165 °C

photocatalyst activation

530 nm

SMILES string

COC(C=C1)=CC=C1C2=CC=C3C4=[N]2[Cu+]5([N]6=C(C7=[N]5C(C8=CC=C(C=C8)OC)=CC=C7C=C9)C9=CC=C6C%10=CC=C(C=C%10)OC)[N]%11=C4C(C=C3)=CC=C%11C%12=CC=C(C=C%12)OC.[Cl-]

InChI

1S/2C26H20N2O2.ClH.Cu/c2*1-29-21-11-5-17(6-12-21)23-15-9-19-3-4-20-10-16-24(28-26(20)25(19)27-23)18-7-13-22(30-2)14-8-18;;/h2*3-16H,1-2H3;1H;/q;;;+1/p-1

InChI key

IBGXDIBAFXUDFP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service