Skip to Content
Merck
All Photos(1)

Key Documents

746193

Sigma-Aldrich

Maleimide-PEG6-succinimidyl ester

Synonym(s):

25-(2,5-Dihydro-2,5-dioxo-1H-pyrrol-1-yl)-23-oxo-4,7,10,13,16,19-hexaoxa-22-azapentacosanoic acid 2,5-dioxo-1-pyrrolidinyl ester, Maleimide-PEG6-NHS ester

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$300.00
250 MG
$370.00

$300.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$300.00
250 MG
$370.00

About This Item

Empirical Formula (Hill Notation):
C26H39N3O13
CAS Number:
Molecular Weight:
601.60
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

$300.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

reaction suitability

reaction type: Pegylations
reagent type: cross-linking reagent

functional group

NHS ester
maleimide

storage temp.

−20°C

SMILES string

O=C(ON1C(CCC1=O)=O)CCOCCOCCOCCOCCOCCOCCNC(CCN2C(C=CC2=O)=O)=O

InChI

1S/C26H39N3O13/c30-21(5-8-28-22(31)1-2-23(28)32)27-7-10-37-12-14-39-16-18-41-20-19-40-17-15-38-13-11-36-9-6-26(35)42-29-24(33)3-4-25(29)34/h1-2H,3-20H2,(H,27,30)

InChI key

AVCPTIUOZBWCKE-UHFFFAOYSA-N

Application

Heterobifunctional crosslinker with an ethylene oxide spacer for linking amine- to sulfhydryl-containing compounds or biomolecules. Maleimide functional group will react with sulfhydryls and the succinimidyl ester group will react with amines. Spacer length is 31.7 angstroms.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service