Skip to Content
Merck
All Photos(1)

Key Documents

746010

Sigma-Aldrich

3-(4-Cyanophenyl)propionic acid

97%

Synonym(s):

3-(p-Cyanophenyl)propionic acid, 4-(2-Carboxyethyl)benzonitrile, 4-Cyanobenzenepropionic acid, 4-Cyanohydrocinnamic acid, [p-Cyanophenyl]-3-propanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$120.00

$120.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$120.00

About This Item

Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$120.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

97%

form

solid

mp

138-143 °C

functional group

carboxylic acid
nitrile

SMILES string

N#Cc1ccc(cc1)CCC(=O)O

InChI

1S/C10H9NO2/c11-7-9-3-1-8(2-4-9)5-6-10(12)13/h1-4H,5-6H2,(H,12,13)

InChI key

RHIGOXBTMPLABF-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service