Skip to Content
Merck
All Photos(1)

Key Documents

745820

Sigma-Aldrich

Difluoro{3-ethyl-5-[1-(4-ethyl-3,5-dimethyl-2H-pyrrol-2-ylidene-N)ethyl]-2,4-dimethyl-1H-pyrrolato-N}boron

98% (HPLC)

Synonym(s):

1,3,5,7,8-Pentamethyl-2,6-diethylpyrromethene-difluoroborate, 2,6-Diethyl-1,3,5,7,8-pentamethylpyrromethene-boron difluoride, 3-Ethyl-5-[1-(4-ethyl-3,5-dimethyl-2H-pyrrol-2-ylidene)ethyl]-2,4-dimethyl-1H-pyrrole, boron complex, Exciton Pyrromethene 567, P 567, Pyrromethene 567

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H25BF2N2
CAS Number:
Molecular Weight:
318.21
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98% (HPLC)

form

solid

mp

208-213 °C

fluorescence

λex 522 nm

SMILES string

CC(C1=C(C)C(CC)=C(C)N1B(F)2F)=C3[N]2=C(C)C(CC)=C3C

InChI

1S/C18H25BF2N2/c1-8-15-10(3)17-12(5)18-11(4)16(9-2)14(7)23(18)19(20,21)22(17)13(15)6/h8-9H2,1-7H3

InChI key

DZSMVBDAUBBZJD-UHFFFAOYSA-N

General description

Difluoro{3-ethyl-5-[1-(4-ethyl-3,5-dimethyl-2H-pyrrol-2-ylidene-N)ethyl]-2,4-dimethyl-1H-pyrrolato-N}boron is a pyrromethene dye that is majorly used in liquid and solid state lasers. Its properties include low intersystem crossing and low triplet-triplet absorption which results in a high fluorescent quantum yields.[1]

Application

BODIPY laser dyes are used to generate fluorescent conjugates of proteins; nucleotides; oligonucleotides and dextrans; and to prepare fluorescent enzyme substrates; fatty acids; phospholipids; lipopolysaccharides; receptor ligands and polystyrene microspheres.

Used for solid-state dye laser devices.
Difluoro{3-ethyl-5-[1-(4-ethyl-3,5-dimethyl-2H-pyrrol-2-ylidene-N)ethyl]-2,4-dimethyl-1H-pyrrolato-N}boron can used in the fabrication of lasers for a variety of applications such as optoelectronics, and high intensity data storage.[1][2][3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leonid M Goldenberg et al.
Advanced materials (Deerfield Beach, Fla.), 24(25), 3339-3343 (2012-06-01)
Distributed feedback (DFB) lasers are produced directly in fluorescent azobenzene-containing materials using a single holographic optical step. Surface relief grating capable of producing images in fluorescence microscopy can be holographically formed in a number of materials.
Pyrromethene-HEMA-based photopolymerizable holographic recording material
Blaya S, et al.
Optics Communications, 228(1-3), 55-61 (2003)
The concentration dependence of the normalized photostability of 1, 3, 5, 7, 8-pentamethyl-2, 6-di-t-butylpyrromethene-difluoroborate complex (PM-597) methanol solutions
Sisk WN and Sanders W
Journal of Photochemistry and Photobiology A: Chemistry, 167(2-3), 185-189 (2004)
Influence of plasmon resonance in silver nanoparticles on the properties of stimulated emission of 1, 3, 5, 7, 8-pentamethyl-2, 6-diethylpyrromethene-difluoroborate molecules in film of porous aluminum oxide
Ibrayev NK and Aimukhanov AK
Optics & Laser technology, 115(2-3), 246-250 (2019)
Daminda Navarathne et al.
Chemical communications (Cambridge, England), 47(44), 12125-12127 (2011-10-15)
An efficient cascade FRET was realized in solid state DNA-CTMA thin films using a three chromophore system without any covalent attachments. The extent of energy transfer from Cm102 to SRh was studied and found to improve eight-fold using the bridging

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service