Skip to Content
Merck
All Photos(1)

Key Documents

745111

Sigma-Aldrich

2-Mercaptoethyl ethyl sulfide Silica

≥45 μm particle size (95%)

Synonym(s):

PhosphonicS SEM26

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$360.00
50 G
$1,240.00

$360.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$360.00
50 G
$1,240.00

About This Item

UNSPSC Code:
12352309
NACRES:
NA.22

$360.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

extent of labeling

0.7-1.3 mmol/g loading

particle size

≥45 μm (95%)

General description

2-Mercaptoethyl ethyl sulfide Silica is a mercaptoalkyl functionalized silica with metal scavenging ability.

Application

Silica Supported Metal Scavenger:
Fe(III), Ni(0), Cu(II), Ru(III), Rh(I), Rh(III), Pd(0), Pd(II), Ag(I), Cd(II), Sn(II), Os, Ir(I), Ir(III), Pt(II), Au(I), Au(III), Hg(II), Pb(II)

Analysis Note

sulfur: detected

Legal Information

Sold in collaboration with PhosphonicS Ltd. for research purposes only.
PhosphonicS is a trademark of PhosphonicS Ltd.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service