Skip to Content
Merck
All Photos(1)

Key Documents

744751

Sigma-Aldrich

14-Azido-3,6,9,12-tetraoxatetradecanoic acid solution

~0.5 M in tert-butyl methyl ether, ≥90% (HPLC)

Synonym(s):

2-{2-{2-[2-(2-Azidoethoxy)ethoxy]ethoxy}ethoxy}acetic acid solution, N33-TEG-COOH

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$138.00

$138.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$138.00

About This Item

Empirical Formula (Hill Notation):
C10H19N3O6
CAS Number:
Molecular Weight:
277.27
Beilstein:
7928855
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

$138.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

≥90% (HPLC)

form

liquid

reaction suitability

reaction type: click chemistry
reagent type: linker

concentration

~0.5 M in tert-butyl methyl ether

impurities

≤0.5% water

density

0.750-0.850 g/mL at 20 °C

functional group

azide
carboxylic acid

storage temp.

2-8°C

SMILES string

OC(COCCOCCOCCOCCN=[N+]=[N-])=O

InChI

1S/C10H19N3O6/c11-13-12-1-2-16-3-4-17-5-6-18-7-8-19-9-10(14)15/h1-9H2,(H,14,15)

InChI key

YHTABJLSZLHRFV-UHFFFAOYSA-N

related product

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-18.4 °F

Flash Point(C)

-28 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service