Skip to Content
Merck
All Photos(1)

Key Documents

744239

Sigma-Aldrich

Triphenylphosphine palladium(II) dichloride phosphaadamantane ethyl Silica

Synonym(s):

PhosphonicS L-PAPd1r

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$381.00

$381.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$381.00

About This Item

UNSPSC Code:
12352103
NACRES:
NA.22

$381.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

extent of labeling

0.010-0.030 mmol/g loading

particle size

≥45 μm (95%)

General description

Triphenylphosphine palladium(II) dichloride phosphaadamantane ethyl Silica is a silica supported heterogeneous palladium catalyst containing a phosphatrioxoadamantane ligand that can be used for Suzuki cross-coupling.[1]

Application

Silica Supported Cross-Coupling Catalyst

Analysis Note

palladium: detected

Legal Information

Sold in collaboration with PhosphonicS Ltd. for research purposes only.
PhosphonicS is a trademark of PhosphonicS Ltd.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guino, M.; et al.
J. Mol. Catal. A: Chem., 293, 25-25 (2008)
Silicaethylphosphatrioxaadamantane?A new support for palladium catalysts and evaluation in Suzuki coupling reactions
Guino M, et al.
J. Mol. Catal. A: Chem., 293(1), 25-30 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service