Skip to Content
Merck
All Photos(1)

Key Documents

744069

Sigma-Aldrich

2-Methyltetrahydrofuran

absolute, stored over molecular sieve

Synonym(s):

2-MeTHF, Tetrahydro-2-methylfuran, Tetrahydrosilvan

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$137.00

$137.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
250 ML
$137.00

About This Item

Empirical Formula (Hill Notation):
C5H10O
CAS Number:
Molecular Weight:
86.13
Beilstein:
102448
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
bp:
78-80 °C (lit.)

$137.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

≥99% (GC)

form

liquid

quality

absolute, stored over molecular sieve

reaction suitability

reaction type: Grignard Reaction

impurities

≤0.002% water

refractive index

n20/D 1.406 (lit.)

bp

78-80 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

CC1CCCO1

InChI

1S/C5H10O/c1-5-3-2-4-6-5/h5H,2-4H2,1H3

InChI key

JWUJQDFVADABEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tony W T Bristow et al.
Journal of the American Society for Mass Spectrometry, 25(10), 1794-1802 (2014-08-12)
For on-line monitoring of chemical reactions (batch or continuous flow), mass spectrometry (MS) can provide data to (1) determine the fate of starting materials and reagents, (2) confirm the presence of the desired product, (3) identify intermediates and impurities, (4)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service