Skip to Content
Merck
All Photos(1)

Key Documents

742678

Sigma-Aldrich

(1R,8S,9s)-Bicyclo[6.1.0]non-4-yn-9-ylmethanol

for Copper-free Click Chemistry

Synonym(s):

(1α,8α,9β)-Bicyclo[6.1.0]non-4-yne-9-methanol, endo-9-Hydroxymethylbicyclo[6.1.0]non-4-yne, BCN-OH

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$383.00

$383.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$383.00

About This Item

Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
Beilstein:
21037713
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$383.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

form

powder

composition

carbon content, 79.96%
hydrogen content, 9.39%

reaction suitability

reaction type: click chemistry
reagent type: linker

functional group

hydroxyl

storage temp.

−20°C

SMILES string

[H][C@@]12[C@@]([C@H]2CO)([H])CCC#CCC1

InChI

1S/C10H14O/c11-7-10-8-5-3-1-2-4-6-9(8)10/h8-11H,3-7H2/t8-,9+,10-

InChI key

NSVXZMGWYBICRW-ILWJIGKKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Alcohol functionalized cyclooctyne derivative. Cyclooctynes are useful in strain-promoted copper-free azide-alkyne cycloaddition reactions. This strained cyclooctyne will react with azide functionalized compounds or biomolecules without the need for a copper catalyst to result in a stable triazole linkage.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xifeng Liu et al.
Biomacromolecules, 20(9), 3352-3365 (2019-08-10)
A new PPF-BCN/hyPCL32-N3 injectable system that can be cross-linked by catalyst-free, strain promoted alkyne-azide cycloaddition (SPAAC) click chemistry was developed for tissue engineering applications. The system consisted of two components: PPF-BCN, poly(propylene fumarate) (PPF) functionalized with (1R,8S,9s)-bicyclo[6.1.0]non-4-yn-9-ylmethanol (BCN-OH), and hyPCL32-N3
Readily accessible bicyclononynes for bioorthogonal labeling and three-dimensional imaging of living cells.
Jan Dommerholt et al.
Angewandte Chemie (International ed. in English), 49(49), 9422-9425 (2010-09-22)

Articles

Copper-free click chemistry is an alternative approach to click chemistry that proceeds at a lower activation barrier and is free of cytotoxic transition metal catalysts.

Explore the principles and applications of click chemistry in drug discovery, highlighting efficient reactions that streamline the synthesis of bioactive compounds.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service