Skip to Content
Merck
All Photos(1)

Key Documents

740624

Sigma-Aldrich

1-(2,3-Epoxypropyl)-2-nitroimidazole

97%

Synonym(s):

1-(Oxiranylmethyl)-2-nitro-1H-imidazole, 1-Oxiranylmethyl-2-nitroimidazole, 2-Nitro-1-(oxiranylmethyl)-1H-imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N3O3
CAS Number:
Molecular Weight:
169.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

solid

mp

50-55 °C

functional group

ether
nitro

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1nccn1CC2CO2

InChI

1S/C6H7N3O3/c10-9(11)6-7-1-2-8(6)3-5-4-12-5/h1-2,5H,3-4H2

InChI key

SYFMSLVOHQZYEB-UHFFFAOYSA-N

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service