Skip to Content
Merck
All Photos(3)

Key Documents

739588

Sigma-Aldrich

Potassium 2-methoxypyrimidin-5-yl-5-trifluoroborate

90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5BF3KN2O
CAS Number:
Molecular Weight:
216.01
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

form

solid

mp

239-254 °C

SMILES string

[K+].COc1ncc(cn1)[B-](F)(F)F

InChI

1S/C5H5BF3N2O.K/c1-12-5-10-2-4(3-11-5)6(7,8)9;/h2-3H,1H3;/q-1;+1

InChI key

ACPITJHSWGPRRF-UHFFFAOYSA-N

Application

Potassium 2-methoxypyrimidin-5-yl-5-trifluoroborate can be used as a substrate in:
  • Metal-free synthesis of biaryls through oxidative electrocoupling reaction.[1]
  • Cross-coupling reactions with unactivated alkyl halides in the presence of a nickel catalyst.[2]
  • Suzuki-Miyaura coupling with aryl and heteroaryl halides using a palladium catalyst.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical Synthesis of Biaryls via Oxidative Intramolecular Coupling of Tetra(hetero)arylborates
Music A, et al.
Journal of the American Chemical Society, 142(9), 4341-4348 (2020)
Arif Music et al.
Journal of the American Chemical Society, 142(9), 4341-4348 (2020-02-11)
We report herein versatile, transition metal-free and additive-free (hetero)aryl-aryl coupling reactions promoted by the oxidative electrocoupling of unsymmetrical tetra(hetero)arylborates (TABs) prepared from ligand-exchange reactions on potassium trifluoroarylborates. Exploiting the power of electrochemical oxidations, this method complements the existing organoboron toolbox.
Nickel-catalyzed cross-coupling of potassium aryl-and heteroaryltrifluoroborates with unactivated alkyl halides
Molander GA, et al.
Organic Letters, 12(24), 5783-5785 (2010)
Scope of the Suzuki- Miyaura cross-coupling reactions of potassium heteroaryltrifluoroborates
Molander GA, et al.
The Journal of Organic Chemistry, 74(3), 973-980 (2009)
Gary A Molander et al.
Organic letters, 12(24), 5783-5785 (2010-11-26)
A method for the cross-coupling of alkyl electrophiles with various potassium aryl- and heteroaryltrifluoroborates has been developed. Nearly stoichiometric amounts of organoboron species could be employed to cross-couple a large variety of challenging heteroaryl nucleophiles. Several functional groups were tolerated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service