Skip to Content
Merck
All Photos(1)

Documents

738727

Sigma-Aldrich

Poly(N-isopropylacrylamide-co-acrylamide)

acrylamide 10 mol %, average Mn 20,000, LCST 34-38 °C

Synonym(s):

Acrylamide copolymer, Poly(NIPAM-co-acrylamide)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11NO)x (C3H5NO)y
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

average Mn 20,000

composition

acrylamide, 10 mol %

transition temp

Tg 144-149 °C

Mw/Mn

2.8

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

By altering the physicochemical properties, smart or intelligent drug delivery systems can be designed to deliver therapeutic molecules on-demand. Learn more about the application of stimuli-responsive materials in drug delivery.

Microparticles in drug delivery: Study on controlling chitosan microparticle size and distribution, exploring encapsulation of BSA and TPP cross-linker.

Biomaterials science integrates smart materials into biological research, requiring a deep understanding of biological systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service