Skip to Content
Merck
All Photos(3)

Key Documents

738247

Sigma-Aldrich

3,4-Bis(hydroxymethyl)furan

98%

Synonym(s):

Furan-3,4-dimethanol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$82.30
5 G
$237.00

$82.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$82.30
5 G
$237.00

About This Item

Empirical Formula (Hill Notation):
C6H8O3
CAS Number:
Molecular Weight:
128.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$82.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

density

1.248 g/mL at 25 °C

functional group

hydroxyl

SMILES string

OCc1cocc1CO

InChI

1S/C6H8O3/c7-1-5-3-9-4-6(5)2-8/h3-4,7-8H,1-2H2

InChI key

RNKXUVJWMOMTHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service