Skip to Content
Merck
All Photos(1)

Key Documents

738093

Sigma-Aldrich

17α-Hydroxyprogesterone-2,3,4-13C3

98 atom % 13C, 98% (CP)

Synonym(s):

17α-Hydroxy-4-pregnene-3,20-dione-2,3,4-13C3, 4-Pregnen-17α-ol-3,20-dione-2,3,4-13C3

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$1,040.00
5 MG
$4,700.00

$1,040.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 MG
$1,040.00
5 MG
$4,700.00

About This Item

Empirical Formula (Hill Notation):
13C3C18H30O3
CAS Number:
Molecular Weight:
333.44
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$1,040.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

98 atom % 13C

Assay

98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+3

storage temp.

−20°C

SMILES string

CC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=[13CH][13C](=O)[13CH2]C[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C21H30O3/c1-13(22)21(24)11-8-18-16-5-4-14-12-15(23)6-9-19(14,2)17(16)7-10-20(18,21)3/h12,16-18,24H,4-11H2,1-3H3/t16-,17+,18+,19+,20+,21+/m1/s1/i6+1,12+1,15+1

InChI key

DBPWSSGDRRHUNT-GGSYSODVSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service